Potassium Iodide-Starch Paper, MilliporeSigma
Supplier: MilliporeSigma
1.09512.0003
EM1.09512.0003CS 10827 USD
EM1.09512.0003
Potassium Iodide-Starch Paper, MilliporeSigma
Test Paper Indicator Paper
Potassium iodine paper is used for the determination of oxidizing agents.