Titripur® Ammonium Buffer Solutions for Complexometry (Ammonium Chloride/Ammonia), MilliporeSigma
Supplier: MilliporeSigma
1.09478.1000
EM1.09478.1000CS
464
USD
EM1.09478.1000
Titripur® Ammonium Buffer Solutions for Complexometry (Ammonium Chloride/Ammonia), MilliporeSigma
Buffers
pH Calibration Buffers
- Hazard Class 8
- Corrosive material
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...