Fluorescent indicator F₂₅₄, Millipore®
Supplier: MilliporeSigma
EMD # 1.09182.0050 replaces and is identical to EMD # 9182-1.
Application: Fluorescent indicator F₂₅₄. CAS 68611-47-2, pH 7.0 (H₂O) suspension.
Physicochemical Information
Density: 4.0 g/cm³ (20 °C)
Melting Point: 1500 °C
pH value: 7.0 (H₂O) suspension
Bulk density: 1450 kg/m³
Solubility: <0.1 g/L
CAS Number:
68611-47-2 |
Specification Test Results
Identity | Conforms |
Fe (iron) | ≤0,01% |
Water | ≤0,3% |
Application test for DC | Conforms |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...