Sodium hydroxide 0.2 mol/l (0.2 N) in aqueous solution, Titripur® volumetric solution, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

M-Clarity™ quality level = MQ100

1.09140.1000
EM1.09140.1000CS 697 USD
EM1.09140.1000
Sodium hydroxide 0.2 mol/l (0.2 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: ∼105 °C (1013 hPa)
Density: 1.02 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Form liquid
Amount-of-substance concentration 0.1990 - 0.2010
Measurement uncertainty 0.0006
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR