Potassium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, Titripur® volumetric solution, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

M-Clarity™ quality level = MQ100

1.09112.1000
EM1.09112.1000CS 251 USD
EM1.09112.1000
Potassium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR