Cerium(IV) sulfate 0.1 mol/l (0.1 N) in aqueous solution, Titripur® volumetric solution, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

1.09092.1000
EM1.09092.1000CS 807 USD
EM1.09092.1000
Cerium(IV) sulfate 0.1 mol/l (0.1 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Cerium (IV) sulphate
Formula: Ce(SO₄)₂
Storage Temperature: Ambient
MDL Number: MFCD00148852
CAS Number: 13590-82-4
EINECS: 237-029-5
UN: 3264
ADR: 8,III
Merck Index: 13,02002

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.06 g/cm3 (20 °C)
pH 0.4 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR