Sulfuric acid 0.5 mol/l (1 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
1.09072.4000
1.09072.9010
1.09072.1000
1.09072.5000
EM1.09072.4000CS
534
USD
EM1.09072.4000
EM1.09072.9010
EM1.09072.1000
EM1.09072.5000
Sulfuric acid 0.5 mol/l (1 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Sulfuric acid
Formula:
H₂SO₄ MW: 98.08 Boiling Pt: 330 Melting Pt: 10.38 Density: 1.01 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 3264 ADR: 8 III |
Specification Test Results
Amount-of-substance concentration | 0.4975 - 0.5025 mol/L |
Form | liquid |
Measurement uncertainty | ± 0.0015 mol/L |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...