Sulfuric acid 0.5 mol/l (1 N) in aqueous solution, Titripur® volumetric solution, Supelco®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

1.09072.4000 1.09072.9010 1.09072.1000 1.09072.5000
EM1.09072.4000CS 534 USD
EM1.09072.4000 EM1.09072.9010 EM1.09072.1000 EM1.09072.5000
Sulfuric acid 0.5 mol/l (1 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 3264
ADR: 8 III

Order Now

Specification Test Results

Amount-of-substance concentration 0.4975 - 0.5025 mol/L
Form liquid
Measurement uncertainty ± 0.0015 mol/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR