Hydrochloric acid 2 mol/l (2 N), Titripur® volumetric solution, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

M-Clarity™ quality level = MQ100

1.09063.1000
EM1.09063.1000CS 191 USD
EM1.09063.1000 EM1.09063.9026
Hydrochloric acid 2 mol/l (2 N), Titripur® volumetric solution, Supelco®
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR