Zinc sulfate heptahydrate 99.5-103.0% (by complexometric analysis), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.08883.9050
EM1.08883.9050EA 5875 USD
EM1.08883.9050
Zinc sulfate heptahydrate 99.5-103.0% (by complexometric analysis), EMSURE® for analysis, Supelco®
Zinc sulphate heptahydrate
Formula: ZnSO₄·7H₂O
Melting Pt: 100 °C
Density: 1.97 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149894
CAS Number: 7446-20-0
EINECS: 231-793-3
UN: 3077
ADR: 9,III
Merck Index: 12,10293

Order Now

Specification Test Results

Solubility 965 g/l (20 °C)
Melting Point 100 °C
Molar Mass 287.54 g/mol
Bulk Density 800 - 1000 kg/m3
Density 1.97 g/cm3 (20 °C)
pH 4 - 6 (50 g/l, H2O, 20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR