Zinc sulfate 0.1 mol/l (0.1 N) in aqueous solution, Titripur® Reag. Ph. Eur. volumetric solution, Supelco®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ100

1.08879.1000
EM1.08879.1000CS 385 USD
EM1.08879.1000
Zinc sulfate 0.1 mol/l (0.1 N) in aqueous solution, Titripur® Reag. Ph. Eur. volumetric solution, Supelco®
Zinc sulphate
Formula: O₄SZn
Storage Temperature: Ambient
MDL Number: MFCD00011302
CAS Number: 7733-02-0
EINECS: 231-793-3
ADR: 9,III

Order Now

Specification Test Results

Form liquid
Amount-of-substance concentration 0.0995 - 0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR