Zinc iodide, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.08828.0100
EM1.08828.0100CS 3970 USD
EM1.08828.0100
Zinc iodide, EMSURE® for analysis, Supelco®
Zinc iodide
Formula: I₂Zn
Boiling Pt: 620 °C (1013 hPa)
Melting Pt: 446 °C
Density: 4.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011299
CAS Number: 10139-47-6
EINECS: 233-396-0
UN: 3077
ADR: 9,III
Merck Index: 12,10269

Order Now

Specification Test Results

Solubility 4500 g/l (20 °C)
Melting Point 446 °C
Molar Mass 319.18 g/mol
Bulk Density 1800 kg/m3
Boiling Point 625 °C (1013 hPa)
Density 4.74 g/cm3 (20 °C)
pH 5.6 (50 g/l, H2O, 20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR