Zinc chloride 98.0-100.5% (ZnCl2, complexometric), EMSURE® analytical reagent, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride

M-Clarity™ quality level = MQ300

1.08816.9025
EM1.08816.9025EA 7208 USD
EM1.08816.9025
Zinc chloride 98.0-100.5% (ZnCl2, complexometric), EMSURE® analytical reagent, Supelco®
Zinc chloride

EMSURE® (formerly GR) is a premium grade of reagents for wet chemical analysis, adhering to ACS, ISO, and/or Reag. Ph.Eur quality specifications.


1.08816.9025 replaces and is the same as ZX0065-20. Part # Change only, Same Pack Size, Same Specifications.
1.08816.9025 is offered as an alternative to ZX0065-7. different size, different packaging, same chem spec.

Formula: ZnCl₂
Boiling Pt: 732 °C (1013 hPa)
Melting Pt: 287…304 °C
Density: 2.93 g/cm3 (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 13,10185

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR