Zinc ≥99.9%, sticks, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

1.08782.0500
EM1.08782.0500CS 13088 USD
EM1.08782.0500
Zinc ≥99.9%, sticks, EMSURE® for analysis, Supelco®
Zinc

Sticks, triangular cross section about 8 mm

Formula: Zn
MW: 65.39 g/mol
MDL Number: MFCD00011291
CAS Number: 7440-66-6

Order Now

Specification Test Results

Assay (complexometric) ≥ 99.9 %
As (Arsenic) ≤ 0.00001 %
Cd (Cadmium) ≤ 0.005 %
Cu (Copper) ≤ 0.002 %
Fe (Iron) ≤ 0.005 %
Pb (Lead) ≤ 0.005 %
Sn (Tin) ≤ 0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR