Hydrogen peroxide 35% stabilized, SAFC®

Supplier: MilliporeSigma
Danger

1.08600.2500
EM1.08600.2500EA 156.87 USD
EM1.08600.2500 EM1.08600.9200
Hydrogen peroxide 35% stabilized, SAFC®
Hydrogen peroxide

Contains 0.015% Na₂H₂P₂O₇, 0.01% H₃PO₄, 0.006% NH₄NO₃, 0.001% Sn as preservatives.


Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
The information provided does not imply the suitability of the product for any particular application. It is customer’s sole responsibility, prior to use, to determine that the product is suitable and permitted for the customer’s intended use and application.

Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: 107 °C (1013 hPa)
Melting Pt: –26 °C
Density: 1.11 g/cm³ (20 °C)
MDL Number: MFCD00011333
CAS Number: 7722-84-1
UN: 2014
ADR: 5.1 (8),II

Order Now

Specification Test Results

Assay (manganometric) 34.5 - 36.0 %
Identity passes test
Appearance of the substance passes test
Free acid (as sulfuric acid) ≤ 0.025 %
Other heavy metals (as Pb) ≤ 0.0002 %
Chloride (Cl) ≤ 0.005 %
Residual solvents (ICH Q3C) ...excluded by production process
Non volatile matter ≤ 0.10 %
Residue on ignition ≤ 0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR