EDTA (ethylenediamine tetraacetic acid), Supelco®

Supplier: MilliporeSigma
Warning

1.08452.0250 1.08452.1000
EM1.08452.0250CS 1600 USD
EM1.08452.0250 EM1.08452.1000
EDTA (ethylenediamine tetraacetic acid), Supelco®
EDTA (Ethylenediamine tetraacetic acid)
Formula: C₁₀H₁₆N₂O₈
Boiling Pt: 614 °C (1013 hPa)
Melting Pt: 220 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 13,03546
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.5 g/l (20 °C)
Melting Point 220 °C (decomposition)
Molar Mass 292.24 g/mol
Bulk Density 600 kg/m3
Vapor Pressure  - 0.013 hPa (20 °C)
Flash Point 100 °C DIN 51758
Density 0.86 g/cm3 (20 °C)
pH 2.5 (10 g/l, H2O, 23 °C) (slurry)
Ignition Point 200 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR