Titriplex® IV solution c (DCTA Na₂) = 0.1 mol/l, Titripur® for metal titration, Supelco®
Supplier: MilliporeSigma
Note: The titer is a correction factor to correct for variations of the volumetric solution, the titration equipment, the temperature and other laboratory conditions. For correct titration results it is recommended to determine a titer with the laboratory specific equipment and under laboratory specific conditions directly after opening a new bottle and at regular time intervals.
Formula:
C₁₄H₂₀N₂Na₂O₈ |
CAS Number:
5786-78-7 |
Specification Test Results
Form | liquid |
Amount-of-substance concentration | 0.0995 - 0.1005 |
Measurement uncertainty | +/- 0.0003 |
Traceability | NIST SRM |
The concentration is determined by volumetric titration and refers to 20°C. | |
The certified value is traceable to a primary standard from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 682 zinc) by means of volumetric standard zinc (Art. 1.02409). | |
This volumetric standard is certified by the accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO 17025. | |
The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%. All measurements are carried out in the laboratory of Merck KGaA, Darmstadt, Germany. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...