EDTA disodium salt dihydrate, Titriplex® III, GR for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.08418.9025 1.08418.0100 1.08418.0250 1.08418.1000 1.08418.5000 1.08418.9025
TXL1084189025QEA 8328.65 USD
TXL1084189025Q EM1.08418.0100 EM1.08418.0250 EM1.08418.1000 EM1.08418.5000 EM1.08418.9025
EDTA disodium salt dihydrate, Titriplex® III, GR for analysis, Supelco®
EDTA disodium salt dihydrate

  • In North America 1.08418.0100 replaces but has a superior specification and a different pack size to EX0539-3
  • In North America 1.08418.0250 replaces but has a superior specification and a different pack size to EX0539-1
  • In North America 1.08418.1000 replaces but has a superior specification and a different pack size to EX0539-1

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Melting Pt: 248 °C (dec., lit.)
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 100 g/l (20 °C)
Melting Point 110 °C Elimination of water of crystallisation
Molar Mass 372.24 g/mol
Bulk Density 700 kg/m3
pH 4 - 5 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR