Titriplex® II (EDTA (Ethylenediamine tetraacetic acid)) 99.4-100.6% (by complexometric analysis) ACS, Reag. Ph. Eur., Supelco®
Supplier: MilliporeSigma
|
Warning
|
1.08417.0100
1.08417.0250
1.08417.1000
EM1.08417.0100CS
4312
USD
EM1.08417.0100
EM1.08417.0250
EM1.08417.1000
EM1.08417.5000
Titriplex® II (EDTA (Ethylenediamine tetraacetic acid)) 99.4-100.6% (by complexometric analysis) ACS, Reag. Ph. Eur., Supelco®
EDTA (Ethylenediamine tetraacetic acid)
Formula:
C₁₀H₁₆N₂O₈ Boiling Pt: 614 °C (1013 hPa) Melting Pt: 220 °C Density: 0.86 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003541 CAS Number: 60-00-4 EINECS: 200-449-4 REACH: 01-2119486399-18 Merck Index: 13,03546 |
|
Specification Test Results
Assay (complexometric) | 99.4 - 100.6 % |
Identity (IR-spectrum) | conforms |
Appearance | White to almost white, fine crystalline to crystalline powder. |
in ammonia insoluble components | ≤ 0.005 % |
Heavy metals (as Pb) | ≤ 10 ppm |
Ca (Calcium) | ≤ 10 ppm |
Fe (Iron) | ≤ 10 ppm |
Mg (Magnesium) | ≤ 5 ppm |
Nitrilotriacetic acid | ≤ 0.1 % |
Sulfated ash | ≤ 0.1 % |
Water (according to Karl Fischer) | ≤ 0.2 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...