Toluene, Uvasol® Spectroscopy, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Methylbenzene, Methacide, Benzene, methyl, Phenylmethane, Methane, phenyl-, Toluol, FM approval, Methylbenzol

1.08331.1000
EM1.08331.1000CS 1059 USD
EM1.08331.1000
Toluene, Uvasol® Spectroscopy, Supelco®
Toluene

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol® solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified. Furthermore the transmittance is specified in accordance with Reag. Ph Eur and ACS.

Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6
Melting Pt: -95
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.52 g/l (20 °C)
Melting Point -95 °C
Molar Mass 92.14 g/mol
Boiling Point 110.6 °C (1013 hPa)
Vapor Pressure 29 hPa (20 °C)
Flash Point 4 °C
Refractive Index 1.4968 (20 °C)
Explosion Limit 1.2 - 8 %(V)
Density 0.87 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 535 °C
Viscosity kinematic 0.7 mm2/s (20 °C)
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR