Toluene ≥99.9% (by GC), EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Methylbenzene, Methacide, Benzene, methyl, Phenylmethane, Methane, phenyl-, Toluol, FM approval, Methylbenzol

1.08325.1000 1.08325.2500 1.08325.2511 1.08325.6010 1.08325.6025 1.08325.4000
EM1.08325.1000CS 608 USD
EM1.08325.1000 EM1.08325.2500 EM1.08325.2511 EM1.08325.6010 EM1.08325.6025 EM1.08325.9190 EM1.08325.4000
Toluene ≥99.9% (by GC), EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Toluene
Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6 (lit.)
Melting Pt: -95 (lit.)
Density: 0.865 g/ml (25°C) (lit.)
Flash Pt: 4
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥ 99.9 %
Identity (IR) conforms
Appearance clear
Color ≤ 10 Hazen
Acidity ≤ 0.0002 meq/g
Alkalinity ≤ 0.0006 meq/g
Density (d 20 °C/20 °C) 0.865 - 0.870
Boiling point 109 - 111 °C
Chloride (Cl) ≤ 0.5 ppm
Sulfate (SO₄) ≤ 1 ppm
Thiophene ≤ 0.0001 %
Sulfur compounds (as S) ≤ 0.003 %
Benzene (GC) ≤ 0.005 %
Readily carbonizable substances conforms
Ag (Silver) ≤ 0.000002 %
Al (Aluminium) ≤ 0.00005 %
As (Arsenic) ≤ 0.000002 %
Au (Gold) ≤ 0.00001 %
B (Boron) ≤ 0.000002 %
Ba (Barium) ≤ 0.00001 %
Be (Beryllium) ≤ 0.000002 %
Bi (Bismuth) ≤ 0.00001 %
Ca (Calcium) ≤ 0.00005 %
Cd (Cadmium) ≤ 0.000005 %
Co (Cobalt) ≤ 0.000002 %
Cr (Chromium) ≤ 0.000002 %
Cu (Copper) ≤ 0.000002 %
Fe (Iron) ≤ 0.00001 %
Ga (Gallium) ≤ 0.000002 %
In (Indium) ≤ 0.000002 %
Li (Lithium) ≤ 0.000002 %
Mg (Magnesium) ≤ 0.00001 %
Mn (Manganese) ≤ 0.000002 %
Mo (Molybdenum) ≤ 0.000005 %
Ni (Nickel) ≤ 0.000002 %
Pb (Lead) ≤ 0.00001 %
Pt (Platinum) ≤ 0.000002 %
Sb (Antimony) ≤ 0.000002 %
Sn (Tin) ≤ 0.00001 %
Ti (Titanium) ≤ 0.000005 %
Tl (Thallium) ≤ 0.000005 %
V (Vanadium) ≤ 0.000005 %
Zn (Zinc) ≤ 0.00001 %
Zr (Zirconium) ≤ 0.000002 %
Evaporation residue ≤ 0.0005 %
Water ≤ 0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR