Tetramethylsilane, MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: TMS

M-Clarity™ quality level = MQ100

1.08183.0100
EM1.08183.0100EA 292 USD
EM1.08183.0100
Tetramethylsilane, MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Tetramethylsilane

MagniSolv™ deuterated solvents provide:
• Reliable results in the NMR-spectra by excellent chemical purity
and highest isotopic enrichment.
• Reliable deuteration degrees.
• Low residual water content.
• Determination of water content
in two ways.

Formula: C₄H₁₂Si
MW: 88.22 g/mol
Boiling Pt: 26.6 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0.642 g/cm³ (20 °C)
Flash Pt: –54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008274
CAS Number: 75-76-3
EINECS: 200-899-1
UN: 2749
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR