Sulfuric acid ≥96% DIN EN ISO 307 calibration solution of viscosity acc. to DIN EN ISO 307, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ300

1.08131.1000 1.08131.2500
EM1.08131.1000CS 518 USD
EM1.08131.1000 EM1.08131.2500
Sulfuric acid ≥96% DIN EN ISO 307 calibration solution of viscosity acc. to DIN EN ISO 307, Supelco®
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Order Now

Specification Test Results

Assay (alkalimetric) 95.85 - 96.15 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR