Tetramethylammonium hydroxide 0.1 mol/L (0.1 N) in 2-propanol/methanol, Titripur® Reag. USP for titration in non-aqueous media, Supelco®

Supplier: MilliporeSigma
Danger

1.08124.1000
EM1.08124.0250CS 4728 USD
EM1.08124.0250 EM1.08124.1000
Tetramethylammonium hydroxide 0.1 mol/L (0.1 N) in 2-propanol/methanol, Titripur® Reag. USP for titration in non-aqueous media, Supelco®
Tetramethylammonium hydroxide
Formula: C₄H₁₃NO
MW: 91.15 g/mol
Density: 0.79 g/cm3 (20 °C)
Flash Pt: 15 °C
MDL Number: MFCD00008280
CAS Number: 75-59-2
UN: 1992
ADR: 3 (6.1),II

Order Now

Specification Test Results

Form liquid
Amount-of-substance concentration 0.0995 - 0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR