Bromocresol green indicator, Supelco®

Supplier: MilliporeSigma

1.08121.0001 1.08121.0005 1.08121.0025
EM1.08121.0001CS 12933 USD
EM1.08121.0001 EM1.08121.0005 EM1.08121.0025
Bromocresol green indicator, Supelco®
Bromocresol green
Formula: C₂₁H₁₄Br₄O₅S
Melting Pt: 217…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00005874
CAS Number: 76-60-8
EINECS: 200-972-8
Merck Index: 13,01370
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12b4d4b3d992f8e86746537bf756af442752c853&vid=553ef1836a5b191a43558f8550e78cf2e424446e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 217 - 218 °C
Molar Mass 698.02 g/mol
Bulk Density 350 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR