MColortest™ Ammonium Test, MilliporeSigma
Supplier: MilliporeSigma
1.08024.0001
EM1.08024.0001EA 135 USD
EM1.08024.0001
MColortest™ Ammonium Test, MilliporeSigma
Environmental Test Kits
The test measures both ammonium ions and dissolved ammonia in e.g
groundwater and surface water, seawater, drinking water and aquarium water. The test kit contains the easy to use colour card and the sliding comparator.