Tin, foil, Thickness ∼ 0.04 mm, Supelco®

Supplier: MilliporeSigma

1.07826.0001
EM1.07826.0001CS 41354 USD
EM1.07826.0001
Tin, foil, Thickness ∼ 0.04 mm, Supelco®
Tin
Formula: Sn
MDL Number: MFCD00133862
CAS Number: 7440-31-5

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 232 °C
Molar Mass 118.71 g/mol
Boiling Point 2260 °C (1013 hPa)
Density 7.3 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR