D-(+)-Sucrose, EMPROVE® ESSENTIAL BP, JP, NF, Ph. Eur., SAFC®

Supplier: MilliporeSigma

1.07653.9029
EM1.07653.9013EA 555.32 USD
EM1.07653.9013 EM1.07653.9053 EM1.07653.9029
D-(+)-Sucrose, EMPROVE® ESSENTIAL BP, JP, NF, Ph. Eur., SAFC®
D(+)-Sucrose
Formula: C₁₂H₂₂O₁₁
Melting Pt: 169…170 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Acidity or alkalinity Passes test
Appearance of solution (50%, water) Passes test
Arsenic (As) 0.0001% max
Calcium (Ca) Passes
Chloride (Cl) 0.0035% max
Color number (Ph. Eur.) 45 max
Dextrins Passes test
Electrical conductivity 35µS/cm max
Endotoxins 250I.U./g max
Heavy metals (as Pb) 0.0005% max
Identity Passes
Lead (Pb) 0.00005% max
Loss on drying (105°C) 0.1% max
Organic volatile impurities (according to NF) Conforms
Other residual solvents (Ph.Eur/USP/ICH) Excluded by manufacturing process
Reducing sugars/invert sugar Passes
Residual solvents (Ph.Eur/USP/ICH) of class 3 0.5% max
Specific rotation [a]20/D (26%, water) +66.3-+67.0°
Sulfate (SO4) 0.005% max
Sulfated ash 0.02% max
Sulfite (SO2) 0.0010% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR