Rubidium chloride, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

1.07615.0025
EM1.07615.0025CS 34481 USD
EM1.07615.0025
Rubidium chloride, EMSURE® for analysis, Supelco®
Rubidium chloride
Formula: RbCl
MDL Number: MFCD00011187
CAS Number: 7791-11-9

Order Now

Specification Test Results

Solubility 910 g/l (20 °C)
Melting Point 715 °C
Molar Mass 120.92 g/mol
Bulk Density 1000 - 1200 kg/m3
Boiling Point 1390 °C
Density 2.76 g/cm3 (20 °C)
pH (50 g/l, H2O, 20 °C) neutral


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR