1-(2-Pyridylazo)-2-naphthol (PAN) indicator for metal titration, Supelco®

Supplier: MilliporeSigma

Synonyms: PAN, 1-(2-Pyridylazo)-2-naphthol

1.07531.0005
EM1.07531.0005CS 10878 USD
EM1.07531.0005
1-(2-Pyridylazo)-2-naphthol (PAN) indicator for metal titration, Supelco®
1-(2-Pyridylazo)-2-naphthol
Formula: C₁₅H₁₁N₃O
MW: 249.27 g/mol
Boiling Pt: 412 °C (1013 hPa)
Melting Pt: 137…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00004071
CAS Number: 85-85-8
EINECS: 201-637-9

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 136 - 141 °C
Molar Mass 249.27 g/mol
Bulk Density 190 kg/m3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR