Lead(II) carbonate, EMSURE® ACS for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Lead carbonat

1.07381.0250
EM1.07381.0250CS 10876.52 USD
EM1.07381.0250
Lead(II) carbonate, EMSURE® ACS for analysis, Supelco®
Lead (II) carbonate
Formula: PbCO₃
MW: 267.21 g/mol
Melting Pt: 315 °C
Density: 6.43 g/cm³ (20 °C)
MDL Number: MFCD00064631
CAS Number: 598-63-0
EINECS: 209-943-4
UN: 2291
ADR: 6.1,III

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 315 °C (decomposition)
Molar Mass 267.21 g/mol
Bulk Density 1300 kg/m3
Density 6.6 g/cm3 (20 °C)
pH 5.8 (50 g/l, H2O, 20 °C) (slurry)
Hazard Class 6.1
Hazard Class Description Toxic materials

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR