Lead(II) acetate trihydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate

1.07375.1000
EM1.07375.1000CS 1462 USD
EM1.07375.1000
Lead(II) acetate trihydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Lead (II) acetate trihydrate
Formula: (H₃CCOO)₂Pb·3H₂O
MW: 379.34 g/mol
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 75 °C
Density: 2.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150023
CAS Number: 6080-56-4
EINECS: 206-104-4
UN: 1616
ADR: 6.1,III
Merck Index: 13,05415
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=351511a44fbb598a2d7fa1fa4ab395427b6168f1&vid=67e858e5f0dc4d04dd796541b3c0ad5aa73f8dc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 443 g/l (20 °C)
Melting Point 75 °C
Molar Mass 379.34 g/mol
Bulk Density 1200 kg/m3
Density 3.3 g/cm3 (20 °C)
pH 5.5 - 6.5 (50 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR