o-Cresolphthalein complexone Reag. Ph. Eur. indicator for metal titration, Supelco®

Supplier: MilliporeSigma

1.07297.0001 1.07297.0010
EM1.07297.0001CS 16602 USD
EM1.07297.0001 EM1.07297.0010
o-Cresolphthalein complexone Reag. Ph. Eur. indicator for metal titration, Supelco®
o-Cresolphthalein complexone
Formula: C₃₂H₃₂N₂O₁₂
Melting Pt: 186 °C
Storage Temperature: Ambient
MDL Number: MFCD00005911
CAS Number: 2411-89-4
EINECS: 219-318-8

Order Now

Specification Test Results

Identity (UV/VIS-Spectrum) passes test
Absorption maximum λmax. (Sodium hydroxide solution 0.1 mol/l) 577 - 580 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; sodium hydroxide solution 0.1 mol/l; calc. on anhydrous substance) ≥ 750
Water (according to Karl Fischer) ≤ 10 %
Suitability as indicator (for metal titration) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR