Palladium(II) nitrate 10.0 ± 0.2 g/L in nitric acid 15% matrix modifier solution, for graphite furnace AAS, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ100
1.07289.0050
EM1.07289.0050CS
16361
USD
EM1.07289.0050
Palladium(II) nitrate 10.0 ± 0.2 g/L in nitric acid 15% matrix modifier solution, for graphite furnace AAS, Supelco®
Palladium (II) nitrate
Palladium matrix modifier for graphite furnace AAS.
Formula:
N₂O₆Pd MW: 230.43 g/mol Density: 1.11 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011169 CAS Number: 10102-05-3 UN: 2031 ADR: 8,II |
Specification Test Results
Assay (palladium) | 9.8 - 10.2 g/l |
Ag (Silver) | ≤ 0.000020 g/l |
As (Arsenic) | ≤ 0.000010 g/l |
Cd (Cadmium) | ≤ 0.000001 g/l |
Pb (Lead) | ≤ 0.000010 g/l |
Se (Selenium) | ≤ 0.00010 g/l |
Tl (Thallium) | ≤ 0.000020 g/l |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...