Phenol red solution indicator, Supelco®

Supplier: MilliporeSigma

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

1.07242.0100
EM1.07242.0100CS 2076 USD
EM1.07242.0100
Phenol red solution indicator, Supelco®
Phenol red
Formula: C₁₉H₁₄O₅S
Density: 0.99 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003552
CAS Number: 143-74-8

Order Now

Specification Test Results

Application test conforms
pH-value (20 °C) 7.0 - 7.3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR