Phenol red indicator, Supelco®

Supplier: MilliporeSigma

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

1.07241.0005 1.07241.0025 1.07241.0100 1.07241.1000
EM1.07241.0005CS 6015 USD
EM1.07241.0005 EM1.07241.0025 EM1.07241.0100 EM1.07241.1000
Phenol red indicator, Supelco®
Phenol red
Formula: C₁₉H₁₄O₅S
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003552
CAS Number: 143-74-8
EINECS: 205-609-7
Merck Index: 13,07329

Order Now

Specification Test Results

Solubility 0.77 g/l (100 °C)
Melting Point 300 °C
Molar Mass 354.38 g/mol
Bulk Density 200 - 300 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR