Phenolphthalein indicator, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
1.07233.0025
1.07233.0100
1.07233.0500
EM1.07233.0025CS
3229
USD
EM1.07233.0025
EM1.07233.0100
EM1.07233.0500
Phenolphthalein indicator, Supelco®
Phenolphthalein
Formula:
C₂₀H₁₄O₄ Melting Pt: 258…262 °C Density: 1.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005913 CAS Number: 77-09-8 EINECS: 201-004-7 REACH: 01-2119498295-24 Merck Index: 14,07243 |
|
Specification Test Results
Solubility | 3.36 mg/l (20 °C) |
Melting Point | 263.7 °C |
Molar Mass | 318.32 g/mol |
Bulk Density | 350 - 450 kg/m3 |
Density | 1.296 g/cm3 (20 °C) |
pH |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...