Phenolphthalein indicator, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

1.07233.0025 1.07233.0100 1.07233.0500
EM1.07233.0025CS 3229 USD
EM1.07233.0025 EM1.07233.0100 EM1.07233.0500
Phenolphthalein indicator, Supelco®
Phenolphthalein
Formula: C₂₀H₁₄O₄
Melting Pt: 258…262 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
REACH: 01-2119498295-24
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 3.36 mg/l (20 °C)
Melting Point 263.7 °C
Molar Mass 318.32 g/mol
Bulk Density 350 - 450 kg/m3
Density 1.296 g/cm3 (20 °C)
pH  - 0.00001 Pa (50 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR