1,10-Phenanthroline monohydrate, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

1.07225.0010 1.07225.0100
EM1.07225.0010CS 27597 USD
EM1.07225.0010 EM1.07225.0100
1,10-Phenanthroline monohydrate, GR ACS, Supelco®
1,10-Phenanthroline monohydrate

1.07225.0010 replaces and is the same as PX0460-5. Part # Change only, Same Pack Size, Same Specifications.

Formula: C₁₂H₈N₂·1H₂O
Melting Pt: 93…94 °C
Density: 1.259 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149973
CAS Number: 5144-89-8
EINECS: 200-629-2
UN: 2811
ADR: 6.1,III
Merck Index: 13,07291
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba346b9f67fdda9afd4d48b893e6318564d604c&vid=2d08e04ee9525293a747b55cdcff8f66a6c4715a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 3.3 g/l (20 °C)
Melting Point 93 - 94 °C
Molar Mass 198.24 g/mol
Bulk Density 300 kg/m3
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR