Paraffin, Solidification point: 56…58 °C, in pastille form for histology, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Wax Paraffin, Paraffin wax

1.07164.2504
EM1.07164.2504EA 295 USD
EM1.07164.2504
Paraffin, Solidification point: 56…58 °C, in pastille form for histology, Sigma-Aldrich®
Paraffin

Paraffin is a traditional embedding media for histology


In order to achieve uniformous ribbon-sectioning quality, defined hardness and homogeneity of the processed material is necessary. This can be guarranteed through high-quality raw materials and defined melting points. The melting point of Paraffin is 56-58°C. It is registered as IVD and CE certified product and can be used for clinical diagnostics.

Density: 0.9 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
CAS Number: 8002-74-2
EINECS: 232-315-6
IVDLogo

Order Now

Specification Test Results

Effect of heating (at 60 °C/100 g) passes test
Solubility in xylene (at 20 °C/0.1 g in 5 ml) passes test
Cutting results (4 μm slices) passes test
Solubility of slices in xylene ≤ 50 s
Solidification point (rotating thermometer method) 56 - 58°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR