Paraffin, viscous Reag. Ph. Eur., Supelco®

Supplier: MilliporeSigma

Synonyms: Wax Paraffin, Paraffin wax

M-Clarity™ quality level = MQ200

1.07160.9026
EM1.07160.9026EA 452 USD
EM1.07160.9026
Paraffin, viscous Reag. Ph. Eur., Supelco®
Paraffin
Boiling Pt: 300…500 °C (1,013 hPa)
Density: 0.86 g/cm3 (20 °C)
Storage Temperature: Ambient
CAS Number: 8002-74-2
EINECS: 232-384-2
Merck Index: 13,07263

Order Now

Specification Test Results

Identity (IR-spectrum) passes test
Identity (Viscosity) passes test
Acidity or alkalinity passes test
Relative Density (d 20/20) 0.827 - 0.890
Dynamic viscosity (20 °C) 110 - 230 mPa∙s
Aromatic, polycyclic Hydrocarbons passes test
Solid Paraffines passes test
Readily carbonisable substance passes test
Sulfur compounds passes test
Corresponds Ph Eur, BP, USP -

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR