8-Quinolinol, GR for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.07098.0250
EM1.07098.0250CS 1943 USD
EM1.07098.0250
8-Quinolinol, GR for analysis, Supelco®
8-Quinolinol
Formula: C₉H₇NO
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 71.2 °C
Density: 1.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006807
CAS Number: 148-24-3
EINECS: 205-711-1
UN: 2811
ADR: 6.1,II
Merck Index: 13,04869
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=752471e0b5caa5507eb8e74e8054dc401912b3ac&vid=ff81328b844178e10beffb1867c97da0d03eda22&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.56 g/l (20 °C)
Melting Point 72.5 - 74.0 °C
Molar Mass 145.16 g/mol
Boiling Point 267 °C (1013 hPa)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR