Anti-static Device, MilliporeSigma
Supplier: MilliporeSigma
1.07070.0001
EM1.07070.0001CS 2064 USD
EM1.07070.0001
Anti-static Device, MilliporeSigma
Surface Treatments Antistatic Treatments
If flammable liquids (e.g
solvents) are to be withdrawn or filled, the container must be properly grounded according to valid safety regulations to avoid explosions and fire risks. The relevant measures are to be taken to discharge static electricity. This can be done by using the antistatic device set. It contains 3 cables for highest flexibility in grounding containers during all work steps. It comes in a suitcase for safe and professional storage of cables that are currently not in use. Cable length: 2 m.