Chloroform, EMPARTA® ACS for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

1.07024.2500
EM1.07024.2500CS 718 USD
EM1.07024.2500
Chloroform, EMPARTA® ACS for analysis, Supelco®
Chloroform
Formula: CHCl₃
MW: 119.38 g/mol
MDL Number: MFCD00000826
CAS Number: 67-66-3
UN: 1888
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 8 g/l (20 °C)
Melting Point -63 °C
Molar Mass 119.38 g/mol
Boiling Point 61 °C (1013 hPa)
Vapor Pressure 211 hPa (20 °C)
Refractive Index 1.45
Density 1.48 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR