Methanol ≥99.8% (by GC), EMPARTA® ACS for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.07018.2511 1.07018.4000 1.07018.9026
EM1.07018.2511CS 371 USD
EM1.07018.2511 EM1.07018.4000 EM1.07018.9026
Methanol ≥99.8% (by GC), EMPARTA® ACS for analysis, Supelco®
Methanol
Formula: CH₃OH
Boiling Pt: 64.7 °C (lit.)
Melting Pt: –98 °C (lit.)
Density: 0.791 g/ml (25°C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -98 °C
Molar Mass 32.04 g/mol
Vapor Pressure 128 hPa (20 °C)
Flash Point 10 °C
Refractive Index 1.33
Explosion Limit 5.5 - 44 %(V)
Density 0.792 g/cm3 (20 °C)
pH 64.5 °C (1013 hPa)
Ignition Point 455 °C DIN 51794
Evaporation Number 6.3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR