Nickel(II) chloride hexahydrate, EMSURE® ACS for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Nickel dichloride hexahydrate

1.06717.0250 1.06717.1000
EM1.06717.0250CS 2046 USD
EM1.06717.0250 EM1.06717.1000
Nickel(II) chloride hexahydrate, EMSURE® ACS for analysis, Supelco®
Nickel (II) chloride hexahydrate
Formula: NiCl₂·6H₂O
Melting Pt: 140 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149809
CAS Number: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 13,06529

Order Now

Specification Test Results

Solubility 553 g/l (20 °C)
Melting Point 1001 °C (anhydrous substance),decomposes
Molar Mass 237.66 g/mol
Bulk Density 640 kg/m3
Boiling Point 987 °C (sublimed),(anhydrous substance)
pH 4.9 (100 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR