Ninhydrin solution in 2-propanol, spray reagent for TLC, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate

M-Clarity™ quality level = MQ100

1.06705.0100
EM1.06705.0100EA 202.6 USD
EM1.06705.0100
Ninhydrin solution in 2-propanol, spray reagent for TLC, Supelco®
Ninhydrin

Ready-to-use spray solution with 2-propanol solvent.

Formula: C₉H₆O₄
MDL Number: MFCD00003791
CAS Number: 485-47-2
UN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ff1a5cd0c772262dac61a6c63dbdeca97baa2c&vid=61d6c7dbd1fe019587f026a878ce0951ce80fcc8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR