Sodium diethyldithiocarbamate trihydrate, GR for analysis, reagent for copper, Supelco®

Supplier: MilliporeSigma
Warning

1.06689.0100
EM1.06689.0100CS 13354 USD
EM1.06689.0100
Sodium diethyldithiocarbamate trihydrate, GR for analysis, reagent for copper, Supelco®
Sodium diethyldithiocarbamate trihydrate
Formula: (C₂H₅)₂NCSSNa·3H₂O
Melting Pt: 95…98.5 °C
Density: 1.1 g/cm³ (20 °C)
MDL Number: MFCD00150617
CAS Number: 20624-25-3
EINECS: 205-710-6
UN: 3077
ADR: 9,III
Merck Index: 12,03443

Order Now

Specification Test Results

Solubility 600 g/l (20 °C)
Melting Point 93 °C
Molar Mass 225.31 g/mol
pH 7 (20 g/l, H2O, 20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous hazardous material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR