Sodium tetraphenylborate, GR for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Tetraphenyl sodium borate, Sodium tetraphenylboron, Tetraphenylboron sodium salt

1.06669.0010 1.06669.0025 1.06669.0100
EM1.06669.0010CS 25149 USD
EM1.06669.0010 EM1.06669.0025 EM1.06669.0100
Sodium tetraphenylborate, GR for analysis, Supelco®
Sodium tetraphenylborate

1.06669.0025 replaces and is the same as SX0805-3. Part # Change only, Same Pack Size, Same Specifications.

Formula: (C₆H₅)₄BNa
Melting Pt: > 400 °C
Density: 1.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011494
CAS Number: 143-66-8
EINECS: 205-605-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,08765

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 300 °C
Molar Mass 342.22 g/mol
Bulk Density 500 kg/m3
pH 8 (50 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR