Sodium sulfate, anhydrous, granulated, EMSURE® for organic trace analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: Disodium sulfate, Bisodium sulfate, Disodium sulphate, Sulfuric acid disodium salt, Bisodium sulphate, Sulphuric acid disodium salt

1.06639.0500
EM1.06639.0500CS 1029 USD
EM1.06639.0500
Sodium sulfate, anhydrous, granulated, EMSURE® for organic trace analysis, Supelco®
Sodium sulphate
Formula: Na₂SO₄
MW: 142.04 g/mol
Boiling Pt: 1429 °C (1013 hPa)
Melting Pt: 888 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance small crystals
Assay (acidimetric) ≥ 99.0 %
Fe (Iron) ≤ 0.001 %
Loss on drying (130 °C) ≤ 0.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR